Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
DL-Aspartic acid
Synonym(s):
(±)-2-Aminosuccinic acid
Linear Formula:
HOOCCH
2
CH(NH
2
)COOH
CAS Number:
617-45-8
Molecular Weight:
133.10
EC Number:
210-513-3
Products
Product Category
amino acids (1)
Brand
Sigma-Aldrich (1)
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Amino Acid
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
A9006
DL-Aspartic acid,
≥99% (TLC)
Expand
View Pricing