Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dithizone
Synonym(s):
Dithizone, Diphenylthiocarbazone
Linear Formula:
C
6
H
5
NHNHCSN=NC
6
H
5
CAS Number:
60-10-6
Molecular Weight:
256.33
EC Number:
200-454-1
Products
Brand
Sigma-Aldrich (2)
Supelco (1)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Markush Class
Markush Group
Special Grade
ACS reagent (1)
Puriss (1)
Puriss P.A. (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
1.03092
Dithizone,
for analysis (1,5-diphenylthiocarbazone) Reag. Ph Eur
Expand
View Pricing
43820
Dithizone,
ACS reagent, for spectrophotometric det. of Cd, Cu, Hg, Pb, Zn, ≥98.0% (TLC)
Expand
View Pricing
D5130
Dithizone,
Practical Grade
Expand
View Pricing