Skip to Content
Merck
Di(propylene glycol) propyl ether, mixture of isomers ≥98.5%

Di(propylene glycol) propyl ether, mixture of isomers

Di(propylene glycol) propyl ether, mixture of isomers ≥98.5%
Linear Formula:
CH3CH2CH2OC3H6OC3H6OH
CAS Number:
Molecular Weight:
176.25
EC Number:

Products

Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Polymer Composition
Polymer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing