
Diethyl sebacate

Synonym(s):
Decanedioic acid 1,10-diethyl ester, NSC 8911, Sebacic acid diethyl ester
Linear Formula:
C2H5OCO(CH2)8COOC2H5
CAS Number:
Molecular Weight:
258.35
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4