Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
D-Asparagine monohydrate
Synonym(s):
(
R
)-(-)-2-Aminosuccinamic acid, (
R
)-2-Aminosuccinic acid 4-amide, (D)-Aspartic acid 4-amide
Linear Formula:
NH
2
COCH
2
CH(NH
2
)COOH · H
2
O
CAS Number:
5794-24-1
Molecular Weight:
150.13
EC Number:
218-163-3
Products
Product Category
amino acids (1)
Brand
Sigma-Aldrich (1)
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
A8131
D-Asparagine monohydrate,
≥99% (TLC)
Expand
View Pricing