
(+)-1-(9-Fluorenyl)ethyl chloroformate solution

Empirical Formula (Hill Notation):
C16H13ClO2
CAS Number:
Molecular Weight:
272.73
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Functional Group
Markush Class
Markush Group
Technique
Showing 1-2 of 2