Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cotton seed GMO standard ERMBF422 for 281-24-236 x 3006-210-23
Products
Manufacturer
Agency
Format
Application
Reference Material Type
Special Grade
certified reference material (4)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
ERMBF422B
Cotton seed GMO standard ERMBF422 for 281-24-236 x 3006-210-23,
ERM
®
, certified reference material, level 1 (98% 281-24-236x3006-210-23 cotton seed), for the detection of GMO cotton seed (281-24-236x3006-210-23) with PCR
Expand
View Pricing
ERMBF422D
Cotton seed GMO standard ERMBF422 for 281-24-236 x 3006-210-23,
ERM
®
, certified reference material, level 3 (10% 281-24-236x3006-210-23 cotton seed), for the detection of GMO cotton seed (281-24-236x3006-210-23) with PCR
Expand
View Pricing
ERMBF422C
Cotton seed GMO standard ERMBF422 for 281-24-236 x 3006-210-23,
ERM
®
, certified reference material, level 2 (1% 281-24-236x3006-210-23 cotton seed), for the detection of GMO cotton seed (281-24-236x3006-210-23) with PCR
Expand
View Pricing
ERMBF422A
Cotton seed GMO standard ERMBF422 for 281-24-236 x 3006-210-23,
ERM
®
, certified reference material, blank (0% 281-24-236x3006-210-23 cotton seed), for the detection of GMO cotton seed (281-24-236x3006-210-23) with PCR
Expand
View Pricing