Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
tissue-culture treated culture dishes
Products
Product Category
dishes (6)
Manufacturer
Diameter (mm)
Surface Area cm²
Package Size
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
CLS430167
Corning
®
tissue-culture treated culture dishes,
D × H 100 mm × 20 mm
Expand
View Pricing
CLS430599
Corning
®
tissue-culture treated culture dishes,
D × H 150 mm × 25 mm
Expand
View Pricing
CLS430165
Corning
®
tissue-culture treated culture dishes,
D × H 35 mm × 10 mm
Expand
View Pricing
CLS430166
Corning
®
tissue-culture treated culture dishes,
D × H 60 mm × 15 mm
Expand
View Pricing
CLS430293
Corning
®
tissue-culture treated culture dishes,
D × H 100 mm × 20 mm
Expand
View Pricing
CLS430196
Corning
®
tissue-culture treated culture dishes,
D × H 60 mm × 15 mm , with 2 mm grid
Expand
View Pricing