Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
LSE
™
rotors for LSE
™
compact centrifuges
Products
Product Category
centrifuges (5)
Manufacturer
Package Size
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
CLS480136
Corning
®
LSE
™
rotors for LSE
™
compact centrifuges,
fixed angle rotor for 6 x 50 mL tubes, 6,000 rpm, 1/cs
Expand
View Pricing
CLS480137
Corning
®
LSE
™
rotors for LSE
™
compact centrifuges,
fixed angle rotor for 12 x 15 mL tubes, 6,000rpm, 1/cs
Expand
View Pricing
CLS480143
Corning
®
LSE
™
rotors for LSE
™
compact centrifuges,
combination rotor for 6 x 15 mL, 2 x 50 mL and 2 x 15 mL
Expand
View Pricing
CLS480138
Corning
®
LSE
™
rotors for LSE
™
compact centrifuges,
swing-out rotor for 6 x 5 mL tubes, 3,500 rpm, 1/cs
Expand
View Pricing
CLS480139
Corning
®
LSE
™
rotors for LSE
™
compact centrifuges,
fixed angle rotor for 18 x 1.5 mL tubes, 6,000rpm, 1/cs
Expand
View Pricing