Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
closed system bottle
Products
Product Category
bottles (6)
Manufacturer
Capacity (mL)
Diameter (mm)
Cap
Color
Vial Bottle Color
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
CLS11666
Corning
®
closed system bottle,
capacity 500 mL, polystyrene, male MPC connector, sterile
Expand
View Pricing
CLS11681
Corning
®
closed system bottle,
capacity 1000 mL, polystyrene, male MPC connector, sterile
Expand
View Pricing
CLS11655
Corning
®
closed system bottle,
capacity 250 mL, polystyrene, male Luer Lock, sterile
Expand
View Pricing
CLS11650
Corning
®
closed system bottle,
capacity 150 mL, polystyrene, male Luer Lock, sterile
Expand
View Pricing
CLS11651
Corning
®
closed system bottle,
capacity 150 mL, polystyrene, male MPC connector, sterile
Expand
View Pricing
CLS11656
Corning
®
closed system bottle,
capacity 250 mL, polystyrene, male MPC connector, sterile
Expand
View Pricing