Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
96 well plates
Products
Product Category
plates (5)
Color
Manufacturer
Binding Type
Diameter (mm)
Volume (µL)
Wells
Color
Membrane Material
Plate Material
Package Size
Sterility
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
CLS3922
Corning
®
96 well plates,
96 well plate, polystyrene, High Bind, white flat bottom wells, non-sterile, white, 100/cs
Expand
View Pricing
CLS3925
Corning
®
96 well plates,
flat bottom, black polystyrene, high binding surface, bag of 25, non-sterile, lid: no
Expand
View Pricing
CLS9102
Corning
®
96 well plates,
Clear Flat Bottom Polystyrene, TC-Treated, Individually Wrapped, Sterile, 1 x 8 Stripwell
™
Expand
View Pricing
CLS3508
Corning
®
96 well plates,
polystyrene plate, clear wells, PVDF membrane non-sterile, case of 50
Expand
View Pricing
CLS4591
Corning
®
96 well plates,
round bottom black, ULA, lid, sterile
Expand
View Pricing