Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Colistin sodium methanesulfonate
Synonym(s):
Colimycin sodium methanesulfonate, Methanesulfonic acid derivative of Polymyxin E, Polymyxin E sodium methanesulfonate
Empirical Formula (Hill Notation):
C
58
H
115
N
16
Na
5
O
28
S
5
CAS Number:
8068-28-8
Molecular Weight:
1759.90
EC Number:
232-516-9
Products
Product Category
antibiotics (2)
Brand
Sigma-Aldrich (2)
Biological Source
Color
Formula Weight
pH Value
Physical Form
Activity Spectrum
Quality Segment
Mode of Action
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
C1511
Colistin sodium methanesulfonate,
~11,500 U/mg
Expand
View Pricing
27655
Colistin sodium methanesulfonate,
from
Bacillus colistinus
Expand
View Pricing