
Cloxacillin benzathine

Empirical Formula (Hill Notation):
C54H56Cl2N8O10S2
CAS Number:
Molecular Weight:
1112.11
Products
Product Category
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing