Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Clonidine hydrochloride
Synonym(s):
2-(2,6-Dichloroanilino)-2-imidazoline hydrochloride
Empirical Formula (Hill Notation):
C
9
H
9
Cl
2
N
3
· HCl
CAS Number:
4205-91-8
Molecular Weight:
266.55
EC Number:
224-121-5
Products
Product Category
bioactive small molecules (1)
Brand
Sigma-Aldrich (1)
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
pharmaceutical primary standard (3)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
C7897
Clonidine hydrochloride,
solid
Expand
View Pricing
1140407
Clonidine hydrochloride,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
C2400000
Clonidine hydrochloride,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing
BP085
Clonidine hydrochloride,
British Pharmacopoeia (BP) Reference Standard
Expand
View Pricing