
Chlordiazepoxide Related Compound A

Synonym(s):
Demoxepam, 7-Chloro-5-phenyl-1,3-dihydro-2H-1,4-benzodiazepin-2-one 4-oxide
Empirical Formula (Hill Notation):
C15H11ClN2O2
CAS Number:
Molecular Weight:
286.71
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing