Skip to Content
Merck
Cerium foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99.9%

Cerium

Cerium foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99.9%
Synonym(s):
Cerium
Empirical Formula (Hill Notation):
Ce
CAS Number:
Molecular Weight:
140.12

Products

Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Diameter (mm)
Length (mm)
Element
Quality Segment
Reagent Type
Core