Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Carbon-
12
C dioxide
Linear Formula:
12
CO
2
CAS Number:
124-38-9
Molecular Weight:
44.00
Products
Product Category
building blocks (2)
gases (2)
stable isotopes (2)
Brand
Sigma-Aldrich (2)
Formula Weight
Melting Point (°C)
Quality Segment
Isotope
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
422606
Carbon-
12
C dioxide,
99.9 atom %
12
C
Expand
View Pricing
600199
Carbon-
12
C dioxide,
99.99 atom %
12
C
Expand
View Pricing