![Calcifediol Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/943/876/08c286a8-8ec3-4ea1-acf9-e0f5668f9edc/640/08c286a8-8ec3-4ea1-acf9-e0f5668f9edc.png)
Calcifediol
![Calcifediol Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/943/876/08c286a8-8ec3-4ea1-acf9-e0f5668f9edc/640/08c286a8-8ec3-4ea1-acf9-e0f5668f9edc.png)
Synonym(s):
25-Hydroxyvitamin D3 monohydrate, 25-Hydroxycholecalciferol
Empirical Formula (Hill Notation):
C27H44O2 · H2O
CAS Number:
Molecular Weight:
418.65
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing