
Calcein

Synonym(s):
Bis[N,N−bis(carboxymethyl)aminomethyl]fluorescein, Fluorescein-bis(methyliminodiacetic acid), Fluorexon
Empirical Formula (Hill Notation):
C30H26N2O13
CAS Number:
Molecular Weight:
622.53
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing