Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
spare burette length for compact automatic burette
Products
Product Category
burettes (8)
Manufacturer
Length (mm)
Volume (ml)
Color
Package Size
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
BR23866
BRAND
®
spare burette length for compact automatic burette,
25 mL, SILBERBRAND
Expand
View Pricing
BR23833
BRAND
®
spare burette length for compact automatic burette,
10 mL, SILBERBRAND
Expand
View Pricing
BR23846
BRAND
®
spare burette length for compact automatic burette,
25 mL, BLAUBRAND
®
Expand
View Pricing
BR23838
BRAND
®
spare burette length for compact automatic burette,
50 mL, SILBERBRAND
Expand
View Pricing
BR23843
BRAND
®
spare burette length for compact automatic burette,
10 mL, BLAUBRAND
®
Expand
View Pricing
BR23836
BRAND
®
spare burette length for compact automatic burette,
25 mL, SILBERBRAND
Expand
View Pricing
BR23868
BRAND
®
spare burette length for compact automatic burette,
50 mL, SILBERBRAND
Expand
View Pricing
BR23848
BRAND
®
spare burette length for compact automatic burette,
50 mL, BLAUBRAND
®
Expand
View Pricing