
Boc-Hyp-OH

Synonym(s):
trans-N-(tert-Butoxycarbonyl)-4-hydroxy-L-proline, Boc-L-hydroxyproline
Empirical Formula (Hill Notation):
C10H17NO5
CAS Number:
Molecular Weight:
231.25
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing