Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Benchmark Incu-Shaker
™
10LR Refrigerated Shaking Incubator with nonslip rubber mat
Synonym(s):
shaker, incubator, incubated shaker, magic clamp, culture
Products
Manufacturer
Length (cm)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
BMSH2012
Benchmark Incu-Shaker
™
10LR Refrigerated Shaking Incubator with nonslip rubber mat,
AC/DC input 115 V
Expand
View Pricing
BMSH2012E
Benchmark Incu-Shaker
™
10LR Refrigerated Shaking Incubator with nonslip rubber mat,
AC/DC input 230 V, EuroPlug
Expand
View Pricing
BMSH2012EUK
Benchmark Incu-Shaker
™
10LR Refrigerated Shaking Incubator with nonslip rubber mat,
AC/DC input 230 V, UK plug
Expand
View Pricing