Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Benchmark Dry Bath Blocks
Synonym(s):
dry bath block, block heater block, drilled block
Products
Product Category
racks (4)
Manufacturer
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
BMSBSWNMR
Benchmark Dry Bath Blocks,
to hold, 30 x NMR tubes
Expand
View Pricing
BMSBSWBCT
Benchmark Dry Bath Blocks,
to hold, blood collection tubes, 12 x 13 mm and 8 x 16mm
Expand
View Pricing
BMSBSW2MTS
Benchmark Dry Bath Blocks,
to hold, microtube strips (2.0 mL) 6 strips
Expand
View Pricing
BMSBSWHEM
Benchmark Dry Bath Blocks,
to hold, 48 X hematocrit tubes, 1.9 mm (not compatible with cover)
Expand
View Pricing