Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Assembled screw cap with hole and TFE/rubber septum
Products
Product Category
septa (4)
Brand
Supelco (3)
Closure Type
Color
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
854987
Assembled screw cap with hole and TFE/rubber septum,
black polypropylene hole cap, thread for 13-425, PTFE/red rubber septum, for use with 4 mL vials with 13-425 thread, pkg of 100 ea
Expand
View Pricing
SU860091
Assembled screw cap with hole and TFE/rubber septum,
black polypropylene hole cap, thread for 8-425, PTFE/rubber septum, for use with 11.6 × 32mm vial (standard opening), pkg of 100 ea
Expand
View Pricing
854984
Assembled screw cap with hole and TFE/rubber septum,
black polypropylene hole cap, thread for 8-425, PTFE/rubber septum, for use with 11.6 × 32 mm vial (standard opening), pkg of 1000 ea
Expand
View Pricing
CB_000233
Assembled screw cap with hole and TFE/rubber septum,
black polypropylene, septum (TEF/red rubber), septum thickness 1.3 mm, 8-425 thread
Expand
View Pricing