Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ammonium metatungstate hydrate
Linear Formula:
(NH
4
)
6
H
2
W
12
O
40
· xH
2
O
CAS Number:
12333-11-8
Molecular Weight:
2956.30 (anhydrous basis)
EC Number:
234-733-4
Products
Brand
Sigma-Aldrich (3)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core
Special Grade
Purum (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
463922
Ammonium metatungstate hydrate,
99.99% trace metals basis
Expand
View Pricing
10035
Ammonium metatungstate hydrate,
≥85% WO
3
basis (gravimetric)
Expand
View Pricing
358975
Ammonium metatungstate hydrate,
≥66.5% (W)
Expand
View Pricing