Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ammonium carbonate
Synonym(s):
Ammonium carbonate
CAS Number:
10361-29-2
Products
Product Category
salts (2)
Brand
SAFC (1)
Supelco (1)
Physical Form
Product Line
Agency
Quality Segment
Application
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
1.59504
Ammonium carbonate,
for analysis EMSURE
®
ACS,Reag. Ph Eur
Expand
View Pricing
1.01136
Ammonium carbonate,
EMPROVE
®
ESSENTIAL, NF
Expand
View Pricing