Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ammonium acetate
Synonym(s):
Ammonium acetate
Linear Formula:
CH
3
CO
2
NH
4
CAS Number:
631-61-8
Molecular Weight:
77.08
EC Number:
211-162-9
Products
Product Category
salts (9)
buffers (7)
Brand
Sigma-Aldrich (9)
Supelco (5)
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
ACS reagent (3)
Puriss (2)
Puriss P.A. (2)
Technique
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
238074
Ammonium acetate,
ACS reagent, ≥97%
Expand
View Pricing
1.01116
Ammonium acetate,
for analysis EMSURE
®
ACS,Reag. Ph Eur
Expand
View Pricing
431311
Ammonium acetate,
≥99.99% trace metals basis
Expand
View Pricing
A1542
Ammonium acetate,
Molecular Biology, ≥98%
Expand
View Pricing
32301
Ammonium acetate,
puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Expand
View Pricing
5.43834
Ammonium acetate,
for HPLC LiChropur
™
Expand
View Pricing
5.33004
Ammonium acetate,
for LC-MS LiChropur
™
Expand
View Pricing
372331
Ammonium acetate,
99.999% trace metals basis
Expand
View Pricing
1.01115
Ammonium acetate,
EMPLURA
®
Expand
View Pricing
73594
Ammonium acetate,
LiChropur
™
, eluent additive for LC-MS
Expand
View Pricing
A7262
Ammonium acetate,
reagent grade, ≥98%
Expand
View Pricing
A7330
Ammonium acetate,
BioXtra, ≥98%
Expand
View Pricing
09689
Ammonium acetate,
BioUltra, ≥99.0%
Expand
View Pricing
32301-M
Ammonium acetate,
puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Expand
View Pricing