Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
α-Methyl-DL-tyrosine methyl ester hydrochloride
Synonym(s):
α-MT methyl ester hydrochloride, 2-Methyl-4-hydroxy-DL-phenylalanine methyl ester hydrochloride, AMPT methyl ester hydrochloride
Linear Formula:
4-(HO)C
6
H
4
CH
2
C(CH
3
)(NH
2
)CO
2
CH
3
· HCl
CAS Number:
7361-31-1
Molecular Weight:
245.70
EC Number:
230-900-0
Products
Product Category
peptides (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
M3281
α-Methyl-DL-tyrosine methyl ester hydrochloride,
≥95% (HPLC)
Expand
View Pricing