Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
α-Mannosidase from
Canavalia ensiformis
(Jack bean)
Synonym(s):
α-D-Mannoside mannohydrolase
CAS Number:
9025-42-7
EC Number:
232-794-1
Products
Product Category
enzymes (1)
Brand
Sigma-Aldrich (1)
Biological Source
Physical Form
Optical Isomer
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
M7257
α-Mannosidase from
Canavalia ensiformis
(Jack bean),
ammonium sulfate suspension, ≥15 units/mg protein (biuret)
Expand
View Pricing