
Aloenin

Synonym(s):
6-[2-(β-D-Glucopyranosyloxy)-4-hydroxy-6-methylphenyl]-4-methoxy-2H-pyran-2-one, Aloearbonaside
Empirical Formula (Hill Notation):
C19H22O10
CAS Number:
Molecular Weight:
410.37
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing