
Alizarin Red S

Synonym(s):
3,4-Dihydroxy-9,10-dioxo-2-anthracenesulfonic acid sodium salt, Alizarin Carmine, Alizarin Red, water soluble, Alizarin sodium monosulfonate, Alizarin sulfonate sodium, Alizarinsulfonic acid sodium salt, Sodium alizarinsulfonate
Empirical Formula (Hill Notation):
C14H7NaO7S
Products
Product Category
Color
Formula Weight
Physical Form
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing