Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Alcian Blue 8GX
Synonym(s):
Alcian Blue, Ingrain Blue 1
Empirical Formula (Hill Notation):
C
56
H
68
Cl
4
CuN
16
S
4
CAS Number:
33864-99-2
Molecular Weight:
1298.86
EC Number:
251-705-7
Products
Product Category
stains (3)
Brand
Sigma-Aldrich (4)
Color
Formula Weight
Physical Form
Product Line
Chemical Composition
Application
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
A5268
Alcian Blue 8GX,
powder
Expand
View Pricing
A3157
Alcian Blue 8GX,
Dye content, ≥50%, certified by the Biological Stain Commission, powder
Expand
View Pricing
05500
Alcian Blue 8GX,
for microscopy (Bact., Bot., Hist.)
Expand
View Pricing
A9186
Alcian Blue 8GX,
BioReagent, suitable for detection of glycoproteins
Expand
View Pricing