Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Adenine 9-β-D-arabinofuranoside
Synonym(s):
9-β-D-Arabinofuranosyladenine, Ara-A, Vidarabine
Empirical Formula (Hill Notation):
C
10
H
13
N
5
O
4
CAS Number:
5536-17-4
Molecular Weight:
267.24
EC Number:
226-893-9
Products
Brand
Sigma-Aldrich (1)
Formula Weight
Gene Alias
Physical Form
Purity
Activity Spectrum
Mode of Action
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
A5762
Adenine 9-β-D-arabinofuranoside,
≥99%
Expand
View Pricing