Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Acetylacetone
Synonym(s):
2,4-Pentanedione
Linear Formula:
CH
3
COCH
2
COCH
3
CAS Number:
123-54-6
Molecular Weight:
100.12
EC Number:
204-634-0
Products
Product Category
solvents (2)
building blocks (1)
Brand
Sigma-Aldrich (3)
Supelco (2)
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
P7754
Acetylacetone,
ReagentPlus
®
, ≥99%
Expand
View Pricing
1.09600
Acetylacetone,
for analysis EMSURE
®
Expand
View Pricing
8.00023
Acetylacetone,
for synthesis
Expand
View Pricing
10916
Acetylacetone,
produced by Wacker Chemie AG, Burghausen, Germany, ≥99.5% (GC)
Expand
View Pricing
05581
Acetylacetone,
analytical standard
Expand
View Pricing