Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
6α-Methylprednisolone 21-hemisuccinate sodium salt
Synonym(s):
6α-Methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione 21-hemisuccinate
Linear Formula:
C
26
H
33
O
8
Na
CAS Number:
2375-03-3
Molecular Weight:
496.53
EC Number:
219-156-8
Products
Product Category
bioactive small molecules (1)
hormones (1)
Biological Source
Formula Weight
Gene Alias
Physical Form
Research Area
Sterility
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
M3781
6α-Methylprednisolone 21-hemisuccinate sodium salt,
lyophilized powder
Expand
View Pricing