Skip to Content
Merck
5-Amino-2-pyridinecarbonitrile 96%

5-Amino-2-pyridinecarbonitrile

5-Amino-2-pyridinecarbonitrile 96%
Synonym(s):
5-Amino-2-cyanopyridine
Empirical Formula (Hill Notation):
C6H5N3
CAS Number:
Molecular Weight:
119.12

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing