Skip to Content
Merck
4-Morpholinecarbonyl chloride 98%

4-Morpholinecarbonyl chloride

4-Morpholinecarbonyl chloride 98%
Empirical Formula (Hill Notation):
C5H8ClNO2
CAS Number:
Molecular Weight:
149.58
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing