
3-Mercaptopropionic acid

Synonym(s):
3-Mercaptopropionic acid
Linear Formula:
HSCH2CH2CO2H
CAS Number:
Molecular Weight:
106.14
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-3 of 3
Compare
Product Number
Description
Pricing