Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
3D Biotek 3D Insert
™
PS scaffold
Products
Product Category
inserts (2)
plates (2)
Color
Manufacturer
Binding Type
Wells
Color
Package Size
Pore size (µm)
Sterility
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
Z724300
3D Biotek 3D Insert
™
PS scaffold,
for 96 well plates, 24 inserts/96 well plate
Expand
View Pricing
Z724181
3D Biotek 3D Insert
™
PS scaffold,
for 12 well plates, 6 inserts
Expand
View Pricing
Z687553
3D Biotek 3D Insert
™
PS scaffold,
for 24 well plates, 12 inserts/24 well plate
Expand
View Pricing
Z687545
3D Biotek 3D Insert
™
PS scaffold,
for 6 well plates, 3 inserts/6 well plate
Expand
View Pricing