
3-Aminobenzyl alcohol

Synonym(s):
3-(Hydroxymethyl)aniline
Linear Formula:
H2NC6H4CH2OH
CAS Number:
Molecular Weight:
123.15
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2