2-Thiopheneacetyl chloride
Synonym(s):
2-Thienylacetyl chloride
Empirical Formula (Hill Notation):
C6H5ClOS
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing