Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2-[(
RS
)-(4-Bromophenyl)phenylmethoxy]-
N
,
N
-dimethylethanamine hydrochloride
Synonym(s):
2-[(4-Bromophenyl)phenylmethoxy]ethyl(dimethyl)ammonium chloride, Bromazine hydrochloride, Bromodiphenhydramine hydrochloride, NSC 36113
Empirical Formula (Hill Notation):
C
17
H
20
BrNO · HCl
CAS Number:
1808-12-4
Molecular Weight:
370.71
EC Number:
217-310-9
Products
Brand
Supelco (1)
Formula Weight
Gene Alias
Purity
Format
Quality Segment
Application
Reference Material Type
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
62881
2-[(
RS
)-(4-Bromophenyl)phenylmethoxy]-
N
,
N
-dimethylethanamine hydrochloride,
pharmaceutical impurity standard
Expand
View Pricing