
(2-Dodecen-1-yl)succinic anhydride

Empirical Formula (Hill Notation):
C16H26O3
Products
Product Category
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing