Skip to Content
Merck
2-Chloro-2,2-difluoroacetophenone 95%

2-Chloro-2,2-difluoroacetophenone

2-Chloro-2,2-difluoroacetophenone 95%
Synonym(s):
α-Chloro-α,α-difluoroacetophenone
Linear Formula:
C6H5COCF2Cl
CAS Number:
Molecular Weight:
190.57

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing