
2-(Bromomethyl)phenylboronic acid

Synonym(s):
α-Bromo-o-tolueneboronic acid, o-Boronobenzyl bromide
Empirical Formula (Hill Notation):
C7H8BBrO2
CAS Number:
Molecular Weight:
214.85
Products
Brand
Formula Weight
Melting Point (°C)
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing