
2-amino-benzothiazol-6-ol

Synonym(s):
2-Amino-6-hydroxybenzothiazole, 2-Amino-6-benzothiazolol, 6-Hydroxy-2-aminobenzothiazole, 6-Hydroxybenzothiazol-2-amine
Empirical Formula (Hill Notation):
C7H6N2OS
CAS Number:
Molecular Weight:
166.20
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing