Skip to Content
Merck
1,4-Cyclohexanedione monoethylene acetal 97%

1,4-Cyclohexanedione monoethylene acetal

1,4-Cyclohexanedione monoethylene acetal 97%
Synonym(s):
1,4-Dioxaspiro[4.5]decan-8-one
Empirical Formula (Hill Notation):
C8H12O3
CAS Number:
Molecular Weight:
156.18

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing