
1,4,7-Trithiacyclononane

Synonym(s):
Triethylene trisulfide
Empirical Formula (Hill Notation):
C6H12S3
CAS Number:
Molecular Weight:
180.35
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing