Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1,2-Distearoyl-
sn
-glycero-3-phosphocholine
Synonym(s):
1,2-Dioctadecanoyl-
sn
-glycero-3-phosphocholine, 3-
sn
-Phosphatidylcholine, 1,2-distearoyl, L-α-Phosphatidylcholine, distearoyl, L-β,γ-Distearoyl-α-lecithin, DSPC, PC(18:0/18:0)
Empirical Formula (Hill Notation):
C
44
H
88
NO
8
P
CAS Number:
816-94-4
Molecular Weight:
790.15
EC Number:
212-440-2
Products
Product Category
glycerides (1)
lipids (1)
metabolites (1)
Brand
Sigma-Aldrich (1)
Biological Source
Formula Weight
Physical Form
Purity
Functional Group
Optical Isomer
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
P1138
1,2-Distearoyl-
sn
-glycero-3-phosphocholine,
≥99%
Expand
View Pricing