
Hyaluronidase from bovine testes

Synonym(s):
Hyaluronate 4-glycanohydrolase, Hyaluronoglucosaminidase, Hyaluronidase, Hyaluronidase, Bovine Testes, Hyaluronidase, Streptomyces hyaluronlyticus nov. sp.
CAS Number:
Products
Brand
Biological Source
Color
Physical Form
Tissue
Quality Segment
Application
Relevant Disease
Technique
Showing 1-10 of 10
Compare
Product Number
Description
Pricing